missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 43/GJA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68678
This item is not returnable.
View return policy
Description
Connexin 43/GJA1 Polyclonal antibody specifically detects Connexin 43/GJA1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Connexin 43/GJA1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GKSDPYHATSGALSPAKDCGSQKYAYFNGCSSPTAPLSPMSPPGYKLVTGDRNNSSCRNYNKQAS | |
| 100 μg | |
| Cancer, Cellular Markers, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 2697 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction