missing translation for 'onlineSavingsMsg'
Learn More

Contactin-3 Antibody (006), Biotin , R&D Systems™

Product Code. 18614600 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18614600 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18614600 Supplier Novus Biologicals Supplier No. NBP289312B
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

Contactin-3 Monoclonal specifically detects Contactin-3 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin-3
Applications ELISA
Classification Monoclonal
Clone 006
Conjugate Biotin
Formulation PBS
Gene Alias contactin 3 (plasmacytoma associated)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Contactin-3 (Met1-Ser1002,708Asp/Asn; NP_065923.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5067
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.