missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
COX-1 Monoclonal antibody specifically detects COX-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
Specifications
| Antigen | COX-1 |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 345305 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, Flow Cytometry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS |
| Host Species | Rat |
| Immunogen | E. coli-derived recombinant human COX-1, Ala24-Leu599, Accession # P23219 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?