missing translation for 'onlineSavingsMsg'
Learn More

COX4 Antibody (CL3515) [Alexa Fluor™ 405], Novus Biologicals™

Product Code. 30524858 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30524858 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30524858 Supplier Novus Biologicals Supplier No. NBP343847AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COX4 Monoclonal antibody specifically detects COX4 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX4
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL3515
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias COX IV-1, COX4-1, COX4COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P13073, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Loading Controls, Neuroendocrinology
Primary or Secondary Primary
Gene ID (Entrez) 1327
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.