missing translation for 'onlineSavingsMsg'
Learn More

CPT1C Antibody (4E4) [Janelia Fluor« 646], Novus Biologicals Biologicals™

Product Code. 30518877 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30518877 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518877 Supplier Novus Biologicals Supplier No. NBP342946JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CPT1C Monoclonal antibody specifically detects CPT1C in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPT1C
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 40000
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias carnitine O-palmitoyltransferase 1, brain isoform, Carnitine O-palmitoyltransferase I, brain isoform, carnitine palmitoyltransferase 1CCPT1-B, carnitine palmitoyltransferase I related C, CATL1, CPT IC, CPT1P, CPTI-B, CPTIC, EC 2.3.1.21, FLJ23809
Host Species Mouse
Immunogen carnitine palmitoyltransferase 1C - (Uniprot# Q8TCG5)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Endocrinology, metabolism, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 126129
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.