missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSK Recombinant Rabbit Monoclonal Antibody (23GB1920), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554222
This item is not returnable.
View return policy
Description
CSK is a tyrosine kinase that has been shown to downregulate the activity of SRC oncoprotein through phosphorylation of its carboxy terminus. Since cell transformation by SRC is caused by various mechanisms that interfere with this phosphorylation, the CSK gene might function as an antioncogene. CSK plays an important role in the regulation of cell growth, differentiation, migration, and immune responses. CSK phosphorylates tyrosine residues located in the C-terminal tails of Src-family kinases including LCK, SRC, HCK, FYN, LYN, CSK, and YES1. CSK is expressed ubiquitously as a protein of approximately 50 KDa in cytosol, but a fraction distributes to lipid rafts due to interaction with the lipid raft-associated protein PAG (Csk-binding protein).
Specifications
| CSK | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CSK | |
| AW212630; CSK; CSK, non-receptor tyrosine kinase; C-SRC kinase; c-src tyrosine kinase; C-terminal Src kinase; MGC117393; p50CSK; protein-tyrosine kinase (CSK); protein-tyrosine kinase CYL; Protein-tyrosine kinase MPK-2; Tyrosine-protein kinase CSK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12988, 1445, 315707 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1920 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P32577, P41240, P41241 | |
| CSK | |
| A synthesized peptide derived from human CSK (50-100AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction