missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSK Recombinant Rabbit Monoclonal Antibody (23GB1920), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | CSK |
|---|---|
| Clone | 23GB1920 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089209
![]() |
Invitrogen™
MA554222 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089179
|
Invitrogen™
MA554223 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSK Recombinant Monoclonal antibody specifically detects CSK in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CSK | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P32577, P41240, P41241 | |
| 12988, 1445, 315707 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1920 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AW212630; CSK; CSK, non-receptor tyrosine kinase; C-SRC kinase; c-src tyrosine kinase; C-terminal Src kinase; MGC117393; p50CSK; protein-tyrosine kinase (CSK); protein-tyrosine kinase CYL; Protein-tyrosine kinase MPK-2; Tyrosine-protein kinase CSK | |
| A synthesized peptide derived from human CSK (50-100AA). | |
| Primary | |
| -20°C | |
| CSK |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title