missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CSRP2BP Polyclonal specifically detects CSRP2BP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | CSRP2BP |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1-4 μg/mL |
| Formulation | PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | ADA2A-containing complex subunit 2, ATAC component 2 homolog, ATAC2CRP2BPKAT14, CRP2 binding partner, CRP2 binding protein, CRP2-binding partner, CSRP2 binding protein, CSRP2-binding protein, cysteine rich protein 2 binding protein, cysteine-rich protein 2-binding protein, dJ717M23.1, MGC15388, PRO1194 |
| Gene Symbols | KAT14 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:IYGAKEGGISRLPAGQATYRTTCQDFRILDRYQTSLPSRKGFRHQTTKFLYRLVGSEDMAVDQSIVSPYTSRILKPYIRRDYETKPPKLQLLSQIRSHLH |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?