missing translation for 'onlineSavingsMsg'
Learn More

CT83 Antibody (CL4762) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30297457 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30297457 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30297457 Supplier Novus Biologicals Supplier No. NBP259062CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CT83 Monoclonal antibody specifically detects CT83 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CT83
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL4762
Conjugate CoraFluor 1
Formulation PBS
Gene Alias chromosome X open reading frame 61, CT83, CT83Cancer/testis antigen 83, CXorf61, FLJ20611, FLJ22913, kita-kyushu lung cancer antigen 1, KKLC1, KK-LC-1, KK-LC-1KKLC1, RP3-452H17.2
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from Q5H943, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203413
Target Species Human
Content And Storage Store at 4⪚C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.