missing translation for 'onlineSavingsMsg'
Learn More

Ctip1 Antibody (BCL11A/123) - BSA Free, Novus Biologicals™

Product Code. p-200057117 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18628606 0.025 mL 0.03mL
18681248 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18628606 Supplier Novus Biologicals Supplier No. NBP2501150.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ctip1 Monoclonal antibody specifically detects Ctip1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ctip1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BCL11A/123
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500
Formulation PBS
Gene Alias B-cell CLL/lymphoma 11A, B-cell CLL/lymphoma 11A (zinc finger protein), B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2, B-cell lymphoma/leukemia 11A, BCL-11A, BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1, BCL11A-S, BCL11A-XL, C2H2-type zinc finger protein, COUP-TF-interacting protein 1, CTIP1ecotropic viral integration site 9, ecotropic viral integration site 9 homolog, Ecotropic viral integration site 9 protein homolog, EVI-9, EVI9FLJ34997, HBFQTL5, KIAA1809BCL11A-L, Zinc finger protein 856, ZNF856FLJ10173
Host Species Mouse
Immunogen C-terminus of extra long isoform of Ctip1, BCL11AXL (aa 637-835)
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Inhibitors Activators and Regulators, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 53335
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.