missing translation for 'onlineSavingsMsg'
Learn More

TCTEX1D1 Antibody, HRP, R&D Systems™

Product Code. 18601931 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18601931 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18601931 Supplier Novus Biologicals Supplier No. NBP281830H
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

TCTEX1D1 Polyclonal antibody specifically detects TCTEX1D1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTEX1D1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias FLJ40873, MGC125768, RP11-266I14.2, Tctex1 domain containing 1
Host Species Rabbit
Immunogen TCTEX1D1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human. The immunogen is located within amino acids 30 - 80 of TCTEX1D1. Amino Acid Squence: SYMEEPSQRDDISRLT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200132
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.