missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CXC-ELR Antibody (Lilly patent anti-Pan-ELR+) - Humanized, IgG4SP, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CXC-ELR Monoclonal antibody specifically detects CXC-ELR in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | CXC-ELR |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | Lilly patent anti-Pan-ELR+ |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| Host Species | Human |
| Immunogen | CXC-ELR |
| Purification Method | Protein A purified |
| Quantity | 1 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?