missing translation for 'onlineSavingsMsg'
Learn More

CXCR7/RDC-1 Antibody, Novus Biologicals™

Product Code. p-200046653 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18473202 25ul 25µL
18036887 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18473202 Supplier Novus Biologicals Supplier No. NBP21388525ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CXCR7/RDC-1 Polyclonal specifically detects CXCR7/RDC-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR7/RDC-1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Gene Symbols ACKR3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: LHLFDYSEPGNFSDISWPCNSSDCIVVDTVMCPNMPNKSVLLY
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 57007
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.