missing translation for 'onlineSavingsMsg'
Learn More

Cyanocobalamin (Vitamin B12) 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30457602
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457602 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457602 Supplier LoGiCal Supplier No. LGCAMP1449.0001
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyanocobalamin
CAS 68-19-9
Analyte or Component Names Cyanocobalamin
Shipping Condition Ice pack (-20°C)
Molecular Formula C63 H88 Co N14 O14 P
Certifications/Compliance ISO 17034
Synonym Cyanocobalamin, Vitamin B12, 5,6-Dimethylbenzimidazolyl cyanocobamide, 5,6-Dimethylbenzimidazolyl-Co-cyanocobamide, Anacobin, Antipernicin, Apikobal, B-Twelve, B-Twelve Ora, Bedodeka, Bedoz, Behepan, Berubi, Berubigen, Betalin 12, Betalin 12 Crystalline, Betaline 12, Betolvex, Byladoce, CN-B12, Cobalamin, cyanide, Cobalamin, cyano-, Cobalamin, cyano-5,6-dimethylbenzimidazole-, Cobalin, Cobamide, cyano-5,6-dimethyl-1H-benzimidazole-, Cobamin, Cotel, Covit, Cromatonbic B12, Crystamin, Crystamine, Cyano-5,6-dimethylbenzimidazolylcobamide, Cyano-B12, Cyanocobalamine, Cycolamin, Cykobemin, Cykobeminet, Cyomin, Cyredin, Cytacon, Cytamen, Cytobion, Depinar, Dicopac Kit, Dobetin, Docemine, Docibin, Docigram, Docivit, Dodecabee, Dodecavite, Dodex, Ducobee, Duodecibin, Eligen B12, Embiol, Emociclina, Eritrone, Erycytol, Erythrotin, Euhaemon, Extrinsic factor, Factor II, Factor II (vitamin), Fresmin, Hemo-b-doze, Hemomin, Hepagon, Hepavis, Hepavit, Hepcovite, LLD factor, Lactobacillus lactis dorner factor, Macrabin, Megalovel, Milbedoce, Millevit, NSC 80365, Nagravon, Nascobal, Normocytin, Optovite B12, Pernaemon, Pernaevit, Pernipuron, Plecyamin, Poyamin, Rebramin, Redamina, Redisol, Regividerm cream, Rheumazin, Rhodacryst, Rubesol, Rubivamine, Rubramin, Rubramin PC, Rubripca, Rubrocitol, SMARTShot B12, Sytobex, Vibalt, Vibisone, Virubra, Vita-rubra, Vitamin B12 preparation, Vitarubin, Vitasprint B12, Vitral, alpha-(5,6-Dimethylbenzimidazol-1-yl)cobamide cyanide, 1H-Benzimidazole, 5,6-dimethyl-1-(3-O-phosphono-alpha-D-ribofuranosyl)-, monoester with cobinamide cyanide, inner salt, Cobamide, alpha-5,6-dimethyl-1H-benzimidazolyl-, cyanide, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1H-benzimidazole, Vitamin B12
InChI Formula InChI=1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57+,59-,60+,61+,62+;;/m1../s1
Solution Type Single Solution
SMILES N#[C-][Co+3]1234[N]5=CN(C=6C=C(C(=CC65)C)C)C7OC(CO)C(OP(=O)([O-])OC(C)CNC(=O)CCC8(C9=C(C%10=[N]4C(=CC%11=[N]3C(=C(C%12=[N]2C(C)(C([N-]91)C8CC(=O)N)C(C)(CC(=O)N)C%12CCC(=O)N)C)C(C)(CC(=O)N)C%11CCC(=O)N)C(C)(C)C%10CCC(=O)N)C)C)C7O
IUPAC Name Cyanocobalamin
Solvent or Matrix Methanol
Industry Type Food Testing
Molecular Weight (g/mol) 1355.37
Quantity 1 mL
Formula Weight 1354.5674 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.