missing translation for 'onlineSavingsMsg'
Learn More

Cyanocobalamin (Vitamin B12), LoGiCal

Product Code. 30460755
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460755 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460755 Supplier LoGiCal Supplier No. LGCFOR1449.00
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Cyanocobalamin (Vitamin B12)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyanocobalamin
CAS 68-19-9
Analyte or Component Names Cyanocobalamin
Shipping Condition Room Temperature
Molecular Formula C63 H88 Co N14 O14 P
Certifications/Compliance ISO 9001
Synonym Cyanocobalamin, Vitamin B12, 5,6-Dimethylbenzimidazolyl cyanocobamide, 5,6-Dimethylbenzimidazolyl-Co-cyanocobamide, Anacobin, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole, Cobinamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-alpha-D-ribofuranosyl-1 H-benzimidazole
InChI Formula InChI=1 S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84 H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57+,59-,60+,61+,62+;;/m1../s1
SMILES N#[C-][Co+3]1234 [N]5=CN(C=6 C=C(C(=CC65)C)C)C7OC(CO)C(OP(=O)([O-])OC(C)CNC(=O)CCC8(C9=C(C%10=[N]4 C(=CC%11=[N]3 C(=C(C%12=[N]2 C(C)(C([N-]91)C8CC(=O)N)C(C)(CC(=O)N)C%12 CCC(=O)N)C)C(C)(CC(=O)N)C%11 CCC(=O)N)C(C)(C)C%10 CCC(=O)N)C)C)C7O
IUPAC Name Cyanocobalamin
Industry Type Forensics
Molecular Weight (g/mol) 1355.37
Quantity 10 mg
Formula Weight 1354.5674 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.