missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99464-100ul
This item is not returnable.
View return policy
Description
Cyclin E2 Polyclonal antibody specifically detects Cyclin E2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Cyclin E2 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| CYCE2, cyclin E2, G1/S-specific cyclin E2, G1/S-specific cyclin-E2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Cyclin E2. | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Cellular Markers | |
| 9134 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction