missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | Cyclin E2 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:300-1:10000 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671776
|
Novus Biologicals
NBP2-99464-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652726
|
Novus Biologicals
NBP2-99464-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin E2 Polyclonal antibody specifically detects Cyclin E2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| Cyclin E2 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Cellular Markers | |
| 0.2 um filtered solution in PBS | |
| 9134 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CYCE2, cyclin E2, G1/S-specific cyclin E2, G1/S-specific cyclin-E2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Cyclin E2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title