missing translation for 'onlineSavingsMsg'
Learn More

Cyclin H Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™

Product Code. 30489702 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30489702 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30489702 Supplier Novus Biologicals Supplier No. NBP335092AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cyclin H Polyclonal antibody specifically detects Cyclin H in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin H
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias CAK, cyclin H, cyclin-dependent kinase-activating kinase complex subunit, cyclin-H, MO15-associated protein, p34CAK complex subunit, p37CDK-activating kinase complex subunit
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human Cyclin H (NP_001230.1).,, Sequence:, MYHNSSQKRHWTFSSEEQLARLRADANRKFRCKAVANGKVLPNDPVFLEPHEEMTLCKYYEKRLLEFCSVFKPAMPRSVVGTACMYFKRFYLNNSVMEYHPRIIMLTCAFLACKVDEFNVSSPQFVGNLRESPLGQEKALEQILEYELLLIQQLNFHLIVHNPYRPFEGFLIDLKTRYPILENPEILRKTADDFLNRIALTDAYLLYTPSQIALTAILSSASRAGITMESYLSESLMLKENRTCLSQLLDIMKSMRNLVKKYEPPRSEEVAVLKQKLERCHSAELALNVITKKRKGYEDDDYVSKKSKHEEEEWTDDDLVESL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 902
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.