missing translation for 'onlineSavingsMsg'
Learn More

Cyclobenzaprine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30460940
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
missing translation for 'unitSize'
1mL
This item is not returnable. View return policy

Product Code. 30460940

missing translation for 'mfr': LoGiCal LGCAMP0062.0201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Cyclobenzaprine Hydrochloride 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclobenzaprine Hydrochloride
CAS 6202-23-9
Analyte or Component Names Cyclobenzaprine Hydrochloride
Shipping Condition Ice Pack (-20°C)
Molecular Formula C20 H21 N . Cl H
Certifications/Compliance ISO 17034
Synonym Amitriptyline Hydrochloride Imp. B (EP), Amitriptyline Imp. B (EP), Nortriptyline Hydrochloride Imp. E (EP), Nortriptyline Imp. E (EP), Cyclobenzaprine Hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride, 10,11 delta-Amitriptyline hydrochloride
InChI Formula InChI=1 S/C20H21N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-14 H,7,15H2,1-2H3;1 H
SMILES Cl.CN(C)CCC=C1c2ccccc2C=Cc3ccccc13
IUPAC Name 3-(dibenzo[2,1-b:1',2'-e][7 ]annulen-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 311.85
Quantity 1 mL
Formula Weight 311.1441 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.