missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cyclophilin B Monoclonal antibody specifically detects Cyclophilin B in Human,Mouse,Rat samples. It is validated for Western Blot,KnockDown
Specifications
Specifications
| Antigen | Cyclophilin B |
| Applications | Western Blot, KnockDown |
| Classification | Monoclonal |
| Clone | CL3915 |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | Cyclophilin B, cyclophilin-like protein, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, Rotamase B, S-cyclophilin, SCYLP |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from P23284, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?