missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide Monohydrate, Mikromol™

Product Code. 18092210
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18092210 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18092210 Supplier Mikromol Supplier No. MM0822.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cyclophosphamide Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclophosphamide Monohydrate
CAS 6055-19-2
Analyte or Component Names Cyclophosphamide Monohydrate
Shipping Condition Room Temperature
Molecular Formula C7 H15 Cl2 N2 O2 P . H2 O
Certifications/Compliance ISO 17034
Synonym Cyclophosphamide monohydrate, 2H-1,3,2-Oxazaphosphorin-2-amine, N,N-bis(2-chloroethyl)tetrahydro-, 2-oxide, monohydrate, 2H-1,3,2-Oxazaphosphorine, 2-[bis(2-chloroethyl)amino]tetrahydro-, 2-oxide, monohydrate, 2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine-2-oxide monohydrate, Cyclophosphamide hydrate, Cyclophosphamide Monohydrate, Endoxan monohydrate
InChI Formula InChI=1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1
IUPAC Name N,N-bis(2-chloroethyl)-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine;hydrate
Molecular Weight (g/mol) 279.1
Quantity 250 mg
Formula Weight 278.0354 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.