missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl phenyl sulfide, 98+%

Product Code. 11368735
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368735 1 g 1g
11378735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368735 Supplier Thermo Scientific Alfa Aesar Supplier No. L12321.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14633-54-6 | C9H10S | 150.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14633-54-6
Molecular Formula C9H10S
Molecular Weight (g/mol) 150.24
MDL Number MFCD00009684
InChI Key YIBKCPJOFAUAKY-UHFFFAOYSA-N
Synonym cyclopropyl phenyl sulfide, cyclopropyl phenyl sulfane, cyclopropylsulfanyl benzene, benzene, cyclopropylthio, cyclopropylphenylsulfide, cyclopropyl phenyl sulphide, unii-72c99lp48p, cyclopropylsulfanyl-benzene, pubchem10736, cyclopropylphenyl sulfide
PubChem CID 123348
IUPAC Name cyclopropylsulfanylbenzene
SMILES C1CC1SC1=CC=CC=C1

Specifications

Density 1.045
Boiling Point 61°C to 62°C (1 mmHg)
Flash Point 91°C (195°F)
Odor Stench
Refractive Index 1.581
Quantity 1 g
UN Number UN3334
Beilstein 2075701
Formula Weight 150.24
Percent Purity ≥98%
Chemical Name or Material Cyclopropyl phenyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.