missing translation for 'onlineSavingsMsg'
Learn More

Cyclothiazide, Dr. Ehrenstorfer

Product Code. 30308802
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30308802 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30308802 Supplier Dr. Ehrenstorfer Supplier No. DREC11836600
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclothiazide
CAS 2259-96-3
Analyte or Component Names Cyclothiazide
Shipping Condition +20°C
Molecular Formula C14 H16 Cl N3 O4 S2
Certifications/Compliance ISO/IEC 17025
Synonym 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 3-bicyclo[2.2.1]hept-5-en-2-yl-6-chloro-3,4-dihydro-, 1,1-dioxide, 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3,4-dihydro-3-(5-norbornen-2-yl)-, 1,1-dioxide (6CI,7CI,8CI), 6-Chloro-3,4-dihydro-3-(5-norbornen-2-yl)-2H-1,2,4-benzothiadiazine-7-sulfonamide-1,1-dioxide, 6-Chloro-3,4-dihydro-3-(5-norbornen-2-yl)-7-sulfamyl-1,2,4-benzothiadiazine-1,1-dioxide, 6-Chloro-3-(5-bicyclo[2.2.1]hept-2-enyl)-7-sulfamoyl-3,4-dihydro-1,2,4-benzothiadiazine 1,1-dioxide, 6-Chloro-3-(5-norbornen-2-yl)-7-sulfamoyl-3,4-dihydro-2H-1,2,4-benzothiadiazine-1,1-dioxide, Anhydron, Aquirel, Cyclothiazide, Doburil, Fluidil, Lilly 35483, MDi 193, Renazide, Valmiran
InChI Formula InChI=1S/C14H16ClN3O4S2/c15-10-5-11-13(6-12(10)23(16,19)20)24(21,22)18-14(17-11)9-4-7-1-2-8(9)3-7/h1-2,5-9,14,17-18H,3-4H2,(H2,16,19,20)
SMILES NS(=O)(=O)c1cc2c(NC(NS2(=O)=O)C3CC4CC3C=C4)cc1Cl
IUPAC Name 3-(5-bicyclo[2.2.1]hept-2-enyl)-6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
Molecular Weight (g/mol) 389.88
Quantity 25 mg
Formula Weight 389.0271 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.