missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cynomolgus Monkey LAG-3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB10395-100
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal specifically detects LAG-3 in Cynomolgus Monkey samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| LAG-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
| Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey LAG-3, Val20-Leu450, Accession # XP_005570011.1 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry, CyTOF | |
| 2561B | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| XP_005570011.1 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3902 | |
| Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu