missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cynomolgus Monkey LAG-3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
208.00 € - 482.00 €
Specifications
| Antigen | LAG-3 |
|---|---|
| Clone | 2561B |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780407
![]() |
R&D Systems
MAB10395-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722537
![]() |
R&D Systems
MAB10395-100 |
100 μg |
482.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAG-3 Monoclonal specifically detects LAG-3 in Cynomolgus Monkey samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| LAG-3 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
| Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey LAG-3, Val20-Leu450, Accession # XP_005570011.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2561B | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Cynomolgus Monkey | |
| XP_005570011.1 | |
| 3902 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title