Learn More
CYP20A1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2A9E2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67433-100UL
Description
The CYP20A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP20A1. This antibody recognizes human, mouse, rat antigen. The CYP20A1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| CYP20A1 | |
| 2A9E2 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| CYP M, CYP20A1, Cytochrome P450 20A1 | |
| 100 μL | |
| 57404 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |