Learn More
CYP20A1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2A9E2, Proteintech
Mouse Monoclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | CYP20A1 |
|---|---|
| Clone | 2A9E2 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18075315
|
Proteintech
CL488-67433-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18047415
|
Proteintech
CL488-67433-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CYP20A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP20A1. This antibody recognizes human, mouse, rat antigen. The CYP20A1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| CYP20A1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 57404 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2A9E2 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| CYP M, CYP20A1, Cytochrome P450 20A1 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.