missing translation for 'onlineSavingsMsg'
Learn More

Cyprodinil, TRC

Product Code. 30133334
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133334 5 g 5g
30133705 100 mg 100mg
17961035 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133334 Supplier Toronto Research Chemicals Supplier No. TRCC9890605G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121552-61-2
Molecular Formula C14 H15 N3
Molecular Weight (g/mol) 225.29
Synonym 4-Cyclopropyl-6-methyl-N-phenyl-2-pyrimidinamine, CGA 219417, CXR 6024, Chorus, Chorus (pesticide), Chorus 75WG, Cyprodinil, Unix, Vangard, Vangard 75WG
IUPAC Name 4-cyclopropyl-6-methyl-N-phenylpyrimidin-2-amine
SMILES Cc1cc(nc(Nc2ccccc2)n1)C3CC3

Specifications

Analyte or Component Names Cyprodinil
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H15N3/c1-10-9-13(11-7-8-11)17-14(15-10)16-12-5-3-2-4-6-12/h2-6,9,11H,7-8H2,1H3,(H,15,16,17)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 5 g
Formula Weight 225.1266
Percent Purity >95%
Chemical Name or Material Cyprodinil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.