missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytarabine EP Impurity A, Mikromol™
Description
Cytarabine EP Impurity A
Specifications
Specifications
| Chemical Name or Material | Uracil Arabinoside |
| CAS | 3083-77-0 |
| Analyte or Component Names | Uracil Arabinoside |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H12 N2 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-β-D-Arabinofuranosylpyrimidine-2,4(1H,3H)-dione (Uracil Arabinoside), Ara-U, Arabinofuranosyluracil, Arabinosyluracil, Arauridine, NSC 68928, Spongouridin, Spongouridine, Uracil arabinoside, Cytarabine Imp. A (EP), 1-beta-d-arabinofuranosylpyrimidine-2,4(1H,3H)-dione (uracil arabonoside), Uracil, 1-beta-D-arabinofuranosyl- (6CI,7CI,8CI), 1-beta-D-Arabinofuranosyluracil, Uracil beta-D-arabinofuranoside, Nelarabine related, Uracil Arabinofuranoside, GR 46541X, 282U85, Cytarabine Impurity A |
| InChI Formula | InChI=1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7+,8-/m1/s1 |
| SMILES | OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N2C=CC(=O)NC2=O |
| IUPAC Name | 1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?