missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome c Antibody (7H8.2C12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 54 publications
Brand: Novus Biologicals NB100-56503
This item is not returnable.
View return policy
Description
Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat, Drosophila, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Cell Surface), Flow (Intracellular).
Specifications
| Cytochrome c | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.05-0.5 ug/ml, Simple Western 2.5 ug/ml, Flow Cytometry 1:20-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Yamasaki et al (2006)), Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Fujimara et al (1998)), Immunoblotting reported in scientific literature (PMID 16436379), Flow (Cell Surface) reported in scientific literature (PMID 16467206), Flow (Intracellular) reported in scientific literature (Mohr et al (2004)) | |
| CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody recognizes total cytochrome C which includes both apocytochrome (i.e. cytochrome in the cytosol without heme attached) and holocytochrome (i.e cytochrome in the mitochondria with heme attached). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 7H8.2C12 | |
| Unconjugated | |
| P99999 | |
| CYCS | |
| Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon CYT were used as the immunogen (Jemmerson et al. 1991). The antibody recognizes an epitope within amino acids 93-104 of pigeon cytochrome C based on competitive ELISA results (Jemmerson et al. 1991). | |
| 0.1 mg | |
| Apoptosis, Cellular Markers, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
| 54205 | |
| Human, Mouse, Rat, Drosophila, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction