missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome P450 2D6 Antibody [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30489610 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30489610 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30489610 Supplier Novus Biologicals Supplier No. NBP335296V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cytochrome P450 2D6 Polyclonal antibody specifically detects Cytochrome P450 2D6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 2D6
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CPD6P450DB1, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, cytochrome P450 2D6, cytochrome P450, family 2, subfamily D, polypeptide 6, cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2, cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2, cytochrome P450, subfamily II (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolising), cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing), Cytochrome P450-DB1, Debrisoquine 4-hydroxylase, EC 1.14.14.1, flavoprotein-linked monooxygenase, -metabolizing)-like 1, MGC120389, MGC120390, microsomal monooxygenase, P450C2D, P450-DB1, polypeptide 6, polypeptide 7 pseudogene 2, polypeptide 8 pseudogene 2, xenobiotic monooxygenase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-230 of human Cytochrome P450 2D6 (NP_000097.3).,, Sequence:, VDLMHRRQRWAARYPPGPLPLPGLGNLLHVDFQNTPYCFDQLRRRFGDVFSLQLAWTPVVVLNGLAAVREALVTHGEDTADRPPVPITQILGFGPRSQGVFLARYGPAWREQRRFSVSTLRNLGLGKKSLEQWVTEEAACLCAAFANHSGRPFRPNGLLDKAVSNVIASLTCGRRFEYDDPRFLRLLDLAQEGLKEESGFLREVLNAVPVL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1565
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.