missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 3A5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92508-0.02ml
This item is not returnable.
View return policy
Description
Cytochrome P450 3A5 Polyclonal antibody specifically detects Cytochrome P450 3A5 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Cytochrome P450 3A5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| aryl hydrocarbon hydroxylase, CP35, CYPIIIA5, cytochrome P-450, cytochrome P450 3A5, Cytochrome P450 HLp2, cytochrome P450, family 3, subfamily A, polypeptide 5, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5, Cytochrome P450-PCN3, DKFZp686L16231, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, niphedipine oxidase, P450PCN3FLJ31317, PCN3, xenobiotic monooxygenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 233-502 of human CYP3A5 (NP_000768.1). FEALNVSLFPKDTINFLSKSVNRMKKSRLNDKQKHRLDFLQLMIDSQNSKETESHKALSDLELAAQSIIFIFAGYETTSSVLSFTLYELATHPDVQQKLQKEIDAVLPNKAPPTYDAVVQMEYLDMVVNETLRLFPVAIRLERTCKKDVEINGVFIPKGSMVVIPTYALHHDPKYWTEPEEFRPERFSKKKDSIDPYIYTPFGTGPRNCIGMRFALMNMKLALIRVLQNFSFKPCKETQIPLKLDTQGLLQPEKPIVLKVDSRDGTLSGE | |
| 0.02 mL | |
| Lipid and Metabolism | |
| 1577 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction