missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 3A5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | Cytochrome P450 3A5 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603612
|
Novus Biologicals
NBP2-92508-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608172
|
Novus Biologicals
NBP2-92508-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome P450 3A5 Polyclonal antibody specifically detects Cytochrome P450 3A5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Cytochrome P450 3A5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 1577 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| aryl hydrocarbon hydroxylase, CP35, CYPIIIA5, cytochrome P-450, cytochrome P450 3A5, Cytochrome P450 HLp2, cytochrome P450, family 3, subfamily A, polypeptide 5, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5, Cytochrome P450-PCN3, DKFZp686L16231, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, niphedipine oxidase, P450PCN3FLJ31317, PCN3, xenobiotic monooxygenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 233-502 of human CYP3A5 (NP_000768.1). FEALNVSLFPKDTINFLSKSVNRMKKSRLNDKQKHRLDFLQLMIDSQNSKETESHKALSDLELAAQSIIFIFAGYETTSSVLSFTLYELATHPDVQQKLQKEIDAVLPNKAPPTYDAVVQMEYLDMVVNETLRLFPVAIRLERTCKKDVEINGVFIPKGSMVVIPTYALHHDPKYWTEPEEFRPERFSKKKDSIDPYIYTPFGTGPRNCIGMRFALMNMKLALIRVLQNFSFKPCKETQIPLKLDTQGLLQPEKPIVLKVDSRDGTLSGE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title