missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D Box Binding Protein Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92224-0.1ml
This item is not returnable.
View return policy
Description
D Box Binding Protein Polyclonal antibody specifically detects D Box Binding Protein in Human samples. It is validated for Western Blot
Specifications
| D Box Binding Protein | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| Albumin D box-binding protein, Albumin D-element-binding protein, D site of albumin promoter (albumin D-box) binding protein, D site-binding protein, DABP, TaxREB302, Tax-responsive enhancer element-binding protein 302 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-325 of human DBP (NP_001343.2). MARPVSDRTPAPLLLGGPAGTPPGGGALLGLRSLLQGTSKPKEPASCLLKEKERKAALPAATTPGPGLETAGPADAPAGAVVGGGSPRGRPGPVPAPGLLAPLLWERTLPFGDVEYVDLDAFLLEHGLPPSPPPPGGPSPEPSPARTPAPSPGPGSCGSASPRSSPGHAPARAALGTASGHRAGLTSRDTPSPVDPDTVEVLMTFEPDPADLALSSIPGHETFDPRRHRFSEEELKPQPIMKKARKIQVPEEQKDEKYWSRRYKNNEAAKRSRDARRLKENQISVRAAFLEKENALLRQEVVAVRQELSHYRAVLSRYQAQHGAL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1628 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction