missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D Box Binding Protein Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | D Box Binding Protein |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638181
|
Novus Biologicals
NBP2-92224-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681411
|
Novus Biologicals
NBP2-92224-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D Box Binding Protein Polyclonal antibody specifically detects D Box Binding Protein in Human samples. It is validated for Western BlotSpecifications
| D Box Binding Protein | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Albumin D box-binding protein, Albumin D-element-binding protein, D site of albumin promoter (albumin D-box) binding protein, D site-binding protein, DABP, TaxREB302, Tax-responsive enhancer element-binding protein 302 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-325 of human DBP (NP_001343.2). MARPVSDRTPAPLLLGGPAGTPPGGGALLGLRSLLQGTSKPKEPASCLLKEKERKAALPAATTPGPGLETAGPADAPAGAVVGGGSPRGRPGPVPAPGLLAPLLWERTLPFGDVEYVDLDAFLLEHGLPPSPPPPGGPSPEPSPARTPAPSPGPGSCGSASPRSSPGHAPARAALGTASGHRAGLTSRDTPSPVDPDTVEVLMTFEPDPADLALSSIPGHETFDPRRHRFSEEELKPQPIMKKARKIQVPEEQKDEKYWSRRYKNNEAAKRSRDARRLKENQISVRAAFLEKENALLRQEVVAVRQELSHYRAVLSRYQAQHGAL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 1628 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title