missing translation for 'onlineSavingsMsg'
Learn More
Learn More
d,l-Pheniramine.maleate, Lipomed
Description
d,l-Pheniramine.maleate
Specifications
Specifications
| Chemical Name or Material | Pheniramine Maleate |
| CAS | 132-20-7 |
| Analyte or Component Names | Pheniramine Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H20 N2 . C4 H4 O4 |
| Synonym | Brompheniramine Maleate Imp. C (EP), Brompheniramine Imp. C (EP), Dexchlorpheniramine Maleate Imp. A (EP), Dexchlorpheniramine Imp. A (EP), (3 RS)-N,N-Dimethyl-3-phenyl-3-(pyridin-2-yl)propan-1-amine Maleate, N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1) |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CN(C)CCC(c1ccccc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N,N-dimethyl-3-phenyl-3-pyridin-2-ylpropan-1-amine |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?