missing translation for 'onlineSavingsMsg'
Learn More

D4-GDI/RhoGDI2 Antibody (5A6) [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30518401 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30518401 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518401 Supplier Novus Biologicals Supplier No. NBP342930MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

D4-GDI/RhoGDI2 Monoclonal antibody specifically detects D4-GDI/RhoGDI2 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen D4-GDI/RhoGDI2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A6
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias ARHGDIB Rho GDP dissociation inhibitor (GDI) beta, D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2
Host Species Mouse
Immunogen Rho GDP dissociation inhibitor(GDI) beta - (Uniprot# P52566)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 397
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.