missing translation for 'onlineSavingsMsg'
Learn More

DAP3 Antibody, FITC, R&D Systems™

Product Code. 18604555 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18604555 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18604555 Supplier Novus Biologicals Supplier No. NBP298784F
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

DAP3 Polyclonal specifically detects DAP3 in Human samples. It is validated for Immunoprecipitation/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunoprecipitation, Western Blot
Dilution Western Blot, Immunoprecipitation
Content And Storage Store at 4C in the dark.
Product Type Antibody
Product Line Novus
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.