missing translation for 'onlineSavingsMsg'
Learn More

Dapsone, Mikromol™

Product Code. 17933446
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17933446 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17933446 Supplier Mikromol Supplier No. MM1932.00
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dapsone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dapsone
CAS 80-08-0
Analyte or Component Names Dapsone
Shipping Condition Room Temperature
Molecular Formula C12 H12 N2 O2 S
Certifications/Compliance ISO 17034
Synonym Sulfacetamide Sodium Imp. D (EP), Sulfacetamide Imp. D (EP), Dapsone, 4,4'-sulfonyldianiline, Benzenamine, 4,4'-sulfonylbis-, Aniline, 4,4'-sulfonyldi- (7CI,8CI), 1,1'-Sulfonylbis[4-aminobenzene], 1358F, 4,4'-DDS, 4,4'-Dapsone, 4,4'-Diaminodiphenyl sulfone, 4,4'-Sulfonylbisaniline, 4,4'-Sulfonylbisbenzamine, 4,4'-Sulfonyldianiline, 4,4'-Sulfonyldiphenylamine, 4,4'-Sulphonyldianiline, 4-(4-Aminophenylsulfonyl)benzenamine, 4-Aminophenyl sulfone, Aczone, Aradur 9664-1, Aradur 976-1, Araldite HT 976, Atrisone, Avlosulfon, Avlosulfone, Bis(4-aminophenyl) sulfone, Bis(p-aminophenyl) sulfone, Croysulfone, DADFS, DADPS, DDS, DDS (pharmaceutical), DSS, Dapson, Di(4-aminophenyl)sulfone, Di(p-aminophenyl) sulfone, Diaphenylsulfon, Diaphenylsulfone, Diphenasone, Diphone, Disulone, Dumitone, Eporal, F 1358, HB-MC-GE 40, HB-MC-GE 45, HT 9664, HT 976, HT 976-1, HY 976, Hardener HT 976, Lapox K 10, Metabolite C, NSC 6091, Novophone, Seikacure S, Servidapson, Sulfona, Sulfona-Mae, Sulphadione, Sumicure S, Udolac, p,p'-Diaminodiphenyl sulfone, p,p'-Sulfonyldianiline, p-Aminophenyl sulfone, 4-Aminophenyl Sulfone, 4,4'-Sulfonyldianiline, Sulfacetamide Sodium Impurity D, Sulfacetamide Impurity D
InChI Formula InChI=1S/C12H12N2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H,13-14H2
SMILES Nc1ccc(cc1)S(=O)(=O)c2ccc(N)cc2
IUPAC Name 4-(4-aminophenyl)sulfonylaniline
Molecular Weight (g/mol) 248.3
Quantity 250 mg
Formula Weight 248.0619 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.