missing translation for 'onlineSavingsMsg'
Learn More

DCI Antibody, Novus Biologicals™

Product Code. 30228282 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30228282 100 μL 100µL
30231999 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30228282 Supplier Novus Biologicals Supplier No. NBP335218100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DCI Polyclonal antibody specifically detects DCI in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCI
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias acetylene-allene isomerase, D2-enoyl-CoA isomerase, D3, DCI3,2-trans-enoyl-CoA isomerase3,2 trans-enoyl-CoA isomerase, Delta(2)-enoyl-CoA isomerase, Delta(3), delta3, delta2-enoyl-CoA isomerase, dodecenoyl-CoA delta isomerase (3,2 trans-enoyl-CoA isomerase), dodecenoyl-CoA isomerase3,2-trans-enoyl-CoA isomerase, mitochondrial, dodecenoyl-Coenzyme A delta isomerase (3,2 trans-enoyl-Coenzyme A isomerase), EC 5.3.3.8, enoyl-CoA delta isomerase 13,2 trans-enoyl-Coenzyme A isomerase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 63-302 of human DCI (NP_001910.2).,, Sequence:, KNPPVNSLSLEFLTELVISLEKLENDKSFRGVILTSDRPGVFSAGLDLTEMCGRSPAHYAGYWKAVQELWLRLYQSNLVLVSAINGACPAGGCLVALTCDYRILADNPRYCIGLNETQLGIIAPFWLKDTLENTIGHRAAERALQLGLLFPPAEALQVGIVDQVVPEEQVQSTALSAIAQWMAIPDHARQLTKAMMRKATASRLVTQRDADVQNFVSFISKDSIQKSLQMYLERLKEEKG
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1632
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.