missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDHD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82967-25ul
This item is not returnable.
View return policy
Description
DDHD2 Polyclonal antibody specifically detects DDHD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DDHD2 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| DDHD domain containing 2, DDHD domain-containing protein 2, EC 3.1.1.-, KIAA0725PA-PLA1 like, phospholipase DDHD2, SAM, WWE and DDHD domain containing 1, SAM, WWE and DDHD domain-containing protein 1, SAMWD1, sec23p-interacting protein p125-like phosphatidic acid-preferring phospholipaseA1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GDKDNKYVPYSESFSQVLEETYMLAVTLDEWKKKLESPNREIIILHNPKLMVHYQPVAGSDDWGSTPTEQGRPRTV | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23259 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction