missing translation for 'onlineSavingsMsg'
Learn More

DDI1 Antibody - BSA Free, Novus Biologicals™

Product Code. 30559626 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30559626 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30559626 Supplier Novus Biologicals Supplier No. NBP348038

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DDI1 Polyclonal antibody specifically detects DDI1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDI1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias DDI1, DNA-damage inducible 1, homolog 1, DDI1, DNA-damage inducible 1, homolog 1 (S. cerevisiae), DNA-damage inducible 1 homolog 1 (S. cerevisiae), DNA-damage inducible protein 1, FLJ36017, protein DDI1 homolog 1
Host Species Rabbit
Immunogen DDI1, DNA-damage inducible 1, homolog 1. (Uniprot# Q8WTU0)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 414301
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.