missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DDX41 Monoclonal specifically detects DDX41 in Human samples. It is validated for Flow Cytometry/Immunocytochemistry/Immunofluorescence/Protein Array/Western Blot.
Specifications
Specifications
| Antigen | DDX41 |
| Applications | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array, Western Blot |
| Classification | Monoclonal |
| Clone | PCRP-DDX41-1B4 |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | ABSDEAD-box protein abstrakt, DEAD (Asp-Glu-Ala-Asp) box polypeptide 41, DEAD box protein 41, DEAD box protein abstrakt homolog, EC 3.6.1, EC 3.6.4.13,2900024F02Rik, MGC8828, probable ATP-dependent RNA helicase DDX41, putative RNA helicase |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human DDX41 protein (Uniprot: Q9UJV9) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?