missing translation for 'onlineSavingsMsg'
Learn More

Deanol, Mikromol™

Product Code. 18050390
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18050390 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18050390 Supplier Mikromol Supplier No. MM1681.02
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Deanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethylethanolamine
CAS 108-01-0
Analyte or Component Names N,N-Dimethylethanolamine
Shipping Condition Room Temperature
Molecular Formula C4 H11 N O
Certifications/Compliance ISO 9001
Synonym 2-(Dimethylamino)ethanol, (2-Hydroxyethyl)dimethylamine, (Dimethylamino)ethanol, (N,N-Dimethylamino)ethanol, 2-(Dimethylamino)ethanol, 2-(Dimethylamino)ethyl alcohol, 2-(N,N-Dimethylamino)ethanol, Amietol M 21, Aminoalcohol 2mabs, Bimanol, DMAE, DMEA, Dabco DMEA, Deanol, DeuAdd MA 95, Dimethol, Dimethyl(2-hydroxyethyl)amine, Dimethyl(hydroxyethyl)amine, Dimethylethanolamine, Dimethylmonoethanolamine, Jeffcat DMEA, Kalpur P, Lupragen N 101, MA 95, N,N-Dimethyl(2-hydroxyethyl)amine, N,N-Dimethyl-2-aminoethanol, N,N-Dimethyl-2-ethanolamine, N,N-Dimethyl-N-(β-hydroxyethyl)amine, N,N-Dimethyl-β-hydroxyethylamine, N,N-Dimethylethanolamine, N-(2-Hydroxyethyl)-N,N-dimethylamine, N-(2-Hydroxyethyl)dimethylamine, NSC 2652, Niax DMEA, Norcholine, PC CAT DMEA, Propamine A, Rexolin, T 80, Tegoamin DMEA, Texacat DME, Texacat DMEA, Thancat DME, β-(Dimethylamino)ethanol, β-Dimethylaminoethyl alcohol, β-Hydroxyethyldimethylamine
InChI Formula InChI=1S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3
SMILES CN(C)CCO
IUPAC Name 2-(dimethylamino)ethanol
Molecular Weight (g/mol) 89.14
Quantity 100 mg
Formula Weight 89.0841 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.