missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decanoyl-Trp-(D-Asn)-Asp-Thr-Gly-Orn-Asp-(D-Ala)-OH Trifluoroacetic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Decanoyl-Trp-(D-Asn)-Asp-Thr-Gly-Orn-Asp-(D-Ala)-OH Trifluoroacetic Acid Salt |
| Analyte or Component Names | Decanoyl-Trp-(D-Asn)-Asp-Thr-Gly-Orn-Asp-(D-Ala)-OH Trifluoroacetic Acid Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C47H71N11O16 • (C2HF3O2) |
| Synonym | Daptomycin Impurity B1-II, (2R,5S,8S,17S)-17-((R)-4-Amino-2-((S)-2-decanamido-3-(1H-indol-3-yl)propanamido)-4-oxobutanamido)-8-(3-aminopropyl)-5-(carboxymethyl)-14-((R)-1-hydroxyethyl)-2-methyl-4,7,10,13,16-pentaoxo-3,6,9,12,15-pentaazanonadecane-1,19-dioic Trifluoroacetic Acid Salt, Decanoyl-Trp-(D-Asn)-Asp-Thr-Gly-Orn-Asp-(D-Ala)-OH Trifluoroacetic Acid Salt |
| InChI Formula | InChI=1S/C47H71N11O16.C2HF3O2/c1-4-5-6-7-8-9-10-17-36(61)54-31(19-27-23-50-29-15-12-11-14-28(27)29)43(69)55-32(20-35(49)60)44(70)57-34(22-39(65)66)45(71)58-40(26(3)59)46(72)51-24-37(62)53-30(16-13-18-48)41(67)56-33(21-38(63)64)42(68)52-25(2)47(73)74;3-2(4,5)1(6)7/h11-12,14-15,23,25-26,30-34,40,50,59H,4-10,13,16-22,24,48H2,1-3H3,(H2,49,60)(H,51,72)(H,52,68)(H,53,62)(H,54,61)(H,55,69)(H,56,67)(H,57,70)(H,58,71)(H,63,64)(H,65,66)(H,73,74);(H,6,7)/t25-,26-,30+,31+,32-,33+,34+,40?;/m1./s1 |
| Solution Type | Neat |
| SMILES | O=C(CCCCCCCCC)N[C@H](C(N[C@H](CC(N)=O)C(N[C@@H](CC(O)=O)C(NC(C(NCC(N[C@@H](CCCN)C(N[C@@H](CC(O)=O)C(N[C@@H](C(O)=O)C)=O)=O)=O)=O)[C@H](O)C)=O)=O)=O)CC1=CNC2=C1C=CC=C2.OC(C(F)(F)F)=O |
| IUPAC Name | (2R,5S,8S,17S)-17-((R)-4-amino-2-((S)-2-decanamido-3-(1H-indol-3-yl)propanamido)-4-oxobutanamido)-8-(3-aminopropyl)-5-(carboxymethyl)-14-((R)-1-hydroxyethyl)-2-methyl-4,7,10,13,16-pentaoxo-3,6,9,12,15-pentaazanonadecanedioic acid 2,2,2-trifluoroacetic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?