missing translation for 'onlineSavingsMsg'
Learn More

delta-Sarcoglycan Antibody (19G8), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18103168 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18103168 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18103168 Supplier Novus Biologicals Supplier No. NBP242662AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

delta-Sarcoglycan Monoclonal specifically detects delta-Sarcoglycan in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen delta-Sarcoglycan
Applications Western Blot, ELISA
Classification Monoclonal
Clone 19G8
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD
Gene Symbols SGCD
Host Species Mouse
Immunogen Recombinant human SGCD (57-289aa) purified from E. coli
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 6444
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.