missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus NS1 protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06439-100ul
This item is not returnable.
View return policy
Description
Dengue Virus NS1 protein Polyclonal antibody specifically detects Dengue Virus NS1 protein in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Dengue Virus NS1 protein | |
| Polyclonal | |
| Western Blot 1:2000-1:5000, ELISA 1:5000-1:10000 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Dengue Virus NS1 protein (Accession#: AAC59275.1; Asp1-Ala352). | |
| 100 μg | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto