missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus NS1 protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
322.00 € - 541.00 €
Specifications
| Antigen | Dengue Virus NS1 protein |
|---|---|
| Dilution | Western Blot 1:2000-1:5000, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694948
|
Novus Biologicals
NBP3-06439-100ul |
100 μg |
541.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654648
|
Novus Biologicals
NBP3-06439-50ul |
50 μg |
322.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dengue Virus NS1 protein Polyclonal antibody specifically detects Dengue Virus NS1 protein in Virus samples. It is validated for Western Blot, ELISASpecifications
| Dengue Virus NS1 protein | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:2000-1:5000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Virus | |
| Produced in rabbits immunized with purified, recombinant Dengue Virus NS1 protein (Accession#: AAC59275.1; Asp1-Ala352). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title