missing translation for 'onlineSavingsMsg'
Learn More

Desipramine hydrochloride, Dr. Ehrenstorfer

Product Code. 30315058
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315058 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315058 Supplier Dr. Ehrenstorfer Supplier No. DREC12170167
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desipramine Hydrochloride
CAS 58-28-6
Analyte or Component Names Desipramine Hydrochloride
Shipping Condition +20°C
Molecular Formula C18 H22 N2 . Cl H
Certifications/Compliance ISO/IEC 17025
Synonym Imipramine Hydrochloride Imp. A (EP), Imipramine Imp. A (EP), Desipramine Hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N-methylpropan-1-amine Hydrochloride, Imipramine Hydrochloride Imp. A (EP) as Hydrochloride, 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N-methyl-, hydrochloride (1:1), 5H-Dibenz[b,f]azepine, 10,11-dihydro-5-[3-(methylamino)propyl]-, monohydrochloride (8CI), 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N-methyl-, monohydrochloride (9CI), 10,11-Dihydro-5-[3-(methylamino)propyl]-5H-dibenz[b,f]azepine monohydrochloride, 5-[3-(Methylamino)propyl]-5H-dibenz[b,f]azepine hydrochloride, DMI hydrochloride, Demethylimipramine hydrochloride, Desimipramine-hydrochloride, Desipramine monohydrochloride, Desmethylimipramine chloride, Desmethylimipramine monohydrochloride, Desmethylimipramine-hydrochloride, EX 4355, G 35020, GMI, JB 8181, N-(gamma-Methylaminopropyl)iminodibenzyl hydrochloride, NSC 114901, Norpramin, Nortrimil, Pertofran, Pertofrane, RMI 9384A, Imipramine Hydrochloride Impurity A as Hydrochloride, Imipramine Impurity A as Hydrochloride
InChI Formula InChI=1S/C18H22N2.ClH/c1-19-13-6-14-20-17-9-4-2-7-15(17)11-12-16-8-3-5-10-18(16)20;/h2-5,7-10,19H,6,11-14H2,1H3;1H
SMILES Cl.CNCCCN1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N-methylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 302.84
Quantity 50 mg
Formula Weight 302.155 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.