missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Antibody (5G11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB100-1643
This item is not returnable.
View return policy
Description
Desmoglein-3 Monoclonal specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot.
Specifications
| Desmoglein-3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:1000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 15588331) | |
| Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody reacts with Desmoglein-3. Does not cross-react with Desmoglein 1 or Desmoglein 2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 5G11 | |
| Unconjugated | |
| P32926 | |
| DSG3 | |
| Human Desmoglein 3 recombinant protein [UniProt# P32926] | |
| 0.1 mL | |
| Cytoskeleton Markers, Extracellular Matrix, Signal Transduction | |
| 1830 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction